Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 58 kDa cytokeratin; CK-5; CK-6A; CK-6B; CK-6D; cytokeratin 6A; cytokeratin 6B; cytokeratin 6C; cytokeratin 6D; Cytokeratin-5; Cytokeratin-6A; Cytokeratin-6B; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K5; K6A; K6B; kamp-keratin derived antimicrobial peptide; KDAMP; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6A, , type II; keratin 6A, type II; keratin 6B, type II; keratin K6h; keratin, epidermal type II, K6A; keratin, epidermal, type II, K6B; Keratin, type II cytoskeletal 5; Keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 6B; Keratin-5; Keratin-6A; Keratin-6B; keratin-6C; keratin-like 1 (a type II keratin sequence); Type-II keratin Kb10; type-II keratin Kb12; Type-II keratin Kb5; Type-II keratin Kb6
Gene Aliases: CK-6B; CK-6C; CK-6E; CK5; CK6A; CK6B; CK6C; CK6D; DDD; DDD1; EBS2; K5; K6A; K6B; K6C; K6D; KRT5; KRT5A; KRT6A; KRT6B; KRT6C; KRT6D; KRTL1; PC2; PC3; PC4
UniProt ID: (Human) P13647, (Human) P02538, (Human) P04259
Entrez Gene ID: (Human) 3852, (Human) 3853, (Human) 3854
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support