Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
This antibody is specifically against KRT7.
Immunogen sequence: MSIHFSSPV FTSRSAAFSG RGAQVRLSSA RPGGLGSSSL YGLGASRPRV AVRSAYGGPV GAGIREVTIN QSLLAPLRLD ADPSLQRVRQ EESEQIKTLN NKFASFIDKV RFLEQQNKLL ETKWTLLQEQ KSAKSSRLPD IFEAQIAGLR GQLEALQVDG GRLEAELRSM QDVVEDFKNK YEDEINRRTA AENEFVVLKK DVDAAYMSKV ELEAKVDALN DEINFLRTLN ETELTELQSQ ISDTSVVLSM DNSRSLDLDG IIAEVKAQYE EMAKCSRAEA EAWYQTKFET LQAQAGKHGD DLRNTRNEIS EMNRAIQRLQ AEIDNIKNQR AKLEAAIAEA EERGELALKD AR (1-351 aa encoded by BC002700)
Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-7; cytokeratin 7; Cytokeratin-7; K7; keratin 7, type II; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; Keratin, type II cytoskeletal 7; Keratin-7; Sarcolectin; type II mesothelial keratin K7; Type-II keratin Kb7
Gene Aliases: CK7; K2C7; K7; KRT7; SCL
UniProt ID: (Human) P08729
Entrez Gene ID: (Human) 3855
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support