Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 8
Immunogen sequence: MSIRVTQKSY KVSTSGPRAF SSRSYTSGPG SRISSSSFSR VGSSNFRGGL GGGYGGASGM GGITAVTVNQ SLLSPLVLEV DPNIQAVRTQ EKEQIKTLNN KFASFIDKVR FLEQQNKMLE TKWSLLQQQK TARSNMDNMF ESYINNLRRQ LETLGQEKLK LEAELGNMQG LVEDFKNKYE DEINKRTEME NEFVLIKKDV DEAYMNKVEL ESRLEGLTDE INFLRQLYEE EIRELQSQIS DTSVVLSMDN SRSLDMDSII AEVKAQYEDI ANRSRAEAES MYQIKYEELQ SLAGKHGDDL RRTKTEISEM NRNISRLQAE IEGLKGQRAS LEAAIADAEQ RGELAIKDAN AKLSELEAAL QRAKQDMARQ LREYQELMNV KLALDIEIAT YRKLLEGEES RLESGMQNMS IHTKTTSGYA GGLSSAYGGL TSPGLSYSLG SSFGSGAGSS SFSRTSSSRA VVVKKIETRD GKLVSESSDV LPK; Positive Samples: 293T, NIH/3T3; Cellular Location: Cytoplasm, Nucleus, Nucleus matrix, nucleoplasm
Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-8; cytokeratin 8; Cytokeratin endo A; Cytokeratin-8; K8; keratin 8, type II; keratin complex 2, basic, gene 8; Keratin, type II cytoskeletal 8; Keratin-8; Type-II keratin Kb8
Gene Aliases: AA960620; AL022697; AU019895; CARD2; CK-8; CK8; CYK8; CYKER; EndoA; K2C8; K8; KO; Krt-2.8; Krt2-8; KRT8
UniProt ID: (Human) P05787, (Mouse) P11679, (Rat) Q10758
Entrez Gene ID: (Human) 3856, (Mouse) 16691, (Rat) 25626
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support