Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytosine aminohydrolase; Cytosine deaminase; Fluorocytosine resistance protein 1; yCD
Gene Aliases: FCY1; YP9499.17; YPR062W
UniProt ID: (Baker's yeast) Q12178
Entrez Gene ID: (Baker's yeast) 856175
Molecular Function:
deaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support