Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-20031 detects D-Dimer in human samples.
Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fibrinogen alpha chain; MGC119422; MGC119423; MGC119425
Gene Aliases: ENSMUSG00000059807; Fga; Fib
UniProt ID: (Mouse) E9PV24
Entrez Gene ID: (Mouse) 14161
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support