Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
The DRD2 Dopamine Receptor is involved in the neuromodulation of appetitive behaviors. DRD2 may be linked to alcoholism and other substance-use disorders. Mutations in DRD2 in mice lead to the suppression of reward behavior with morphine and to the reduction of spontaneous movements, a phenotype resembling Parkinsons disease. Three alternatively spliced isoforms have been identified. DRD2 has been reported to be expressed in various regions of the brain, as well as in aorta, artery, eye, and spinal cord. ESTs have been isolated from normal pituitary and in brain, colon, lung, pancreas, and skeletal muscle cancer libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D(2) dopamine receptor; D2 receptor; Dopamine 2 receptor; Dopamine D2 receptor; dopamine receptor 2; dopamine receptor D2 isoform; seven transmembrane helix receptor
Gene Aliases: D2DR; D2R; Drd-2; DRD2
UniProt ID: (Human) P14416, (Mouse) P61168, (Rat) P61169
Entrez Gene ID: (Human) 1813, (Mouse) 13489, (Rat) 24318
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support