Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This gene encodes the D4 subtype of the dopamine receptor. The D4 subtype is a G-protein coupled receptor which inhibits adenylyl cyclase. It is a target for drugs which treat schizophrenia and Parkinson disease. Mutations in this gene have been associated with various behavioral phenotypes, including autonomic nervous system dysfunction, attention deficit/hyperactivity disorder, and the personality trait of novelty seeking. This gene contains a polymorphic number (2-10 copies) of tandem 48 nt repeats; the sequence shown contains four repeats.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D(2C) dopamine receptor; D(4) dopamine receptor; Dopamine 4 receptor; Dopamine D4 receptor; dopamine receptor 4; seven transmembrane helix receptor
Gene Aliases: AW125663; D4DR; D4R; Drd-4; DRD4
UniProt ID: (Human) P21917, (Mouse) P51436
Entrez Gene ID: (Human) 1815, (Mouse) 13491
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support