Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVKLGNNFAE KGTKQPLLED GFDTIPLMTP LDVNQLQFPP PDKVVVKTKT EYEPDRKKGK ARPPQIAEFT VSITEGVTER FKVSVLVLFA LAFLTCVVFL VVYKVYKYDR ACPDGFVLKN TQCIPEGLES YYAEQDSSAR EKFYTVINHY NLAKQSITRS VSPWMSVLSE EKLSEQETEA AEKSA
NEEP21 (neuron-enriched endosomal 21kDa protein), also known as brain neuron cytoplasmic protein 1, NSG1 (neuron-specific proteins family member 1), P21 or D4S234E, is a single pass type II membrane protein belonging to the NSG family. It is highly expressed during neuronal maturation but its expression is downregulated in adult tissues. NEEP21 predominantly localizes to Rab 4-positive early endosomes in the somatodendritic neuronal compartment and is essential for proper receptor sorting and recycling in neurons. It associates with GRIP1 and GluR-2 and mediates the surface expression of GluR-2. When this interaction is interrupted, GluR-2 accumulates in early endosomes and leads to changes in evoked synaptic current properties. In addition, NEEP21 forms a complex with the SNARE protein, Syntaxin 13 (also known as Syntaxin 12), and participates in the recycling of transferrin receptors (TFRs) and NTR2 (Neurotensin receptor 2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain neuron cytoplasmic protein 1; carboxyterminally EE-tagged neuron-enriched endosomal 21 kDa protein; Neuron-enriched endosomal protein of 21 kDa; Neuron-specific protein family member 1; Neuronal vesicle trafficking-associated protein 1
Gene Aliases: D4S234; D4S234E; NEEP21; NSG1; P21
UniProt ID: (Human) P42857
Entrez Gene ID: (Human) 27065
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support