Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIP-1; ASK-interacting protein 1; ASK1-interacting protein 1; DAB2 interaction protein; DAB2-interacting protein; DIP1/2; disabled 2 interacting protein long form; disabled homolog 2 interacting protein; Disabled homolog 2-interacting protein; DOC-2/DAB-2 interactive protein; DOC-2/DAB2 interactive protein; DOC2/DAB2 interactive protein; nGAP-like protein
Gene Aliases: 2310011D08Rik; AF9Q34; AI480459; AIP-1; AIP1; DAB2IP; DIP1/2; KIAA1743; mKIAA1743
UniProt ID: (Human) Q5VWQ8, (Rat) Q6P730, (Mouse) Q3UHC7
Entrez Gene ID: (Human) 153090, (Rat) 192126, (Mouse) 69601
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support