Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is a mouse monoclonal antibody raised against residues near the C terminus of human DACH1.
Immunogen sequence: QHGADSENG DMNSSVGLEL PFMMMPHPLI PVSLPPASVT MAMSQMNHLS TIANMAAAAQ VQSPPSRVET SVIKERVPDS PSPAPSLEEG RRPGSHPSSH RSSSVSSSPA RTESSSDRIP VHQNGLSMNQ MLMGLSPNVL PGPKEGDLAG HDMGHESKRM HIEKDETPLS TPTARDSLDK LSLTGHGQPL PPGFPSPFLF PDGLSSIETL LTNIQGLLKV AIDNARAQEK QVQLEKTELK MDFLRERELR ETLEKQLAME QKNRAIVQKR LKKEKKAKRK LQEALEFETK RREQAEQTLK QAASTDSLRV LNDSLTPEIE ADRSGGRTDA ERTIQDGRLY LKTTVMY (176-521 aa encoded by BC021219 )
Transcription factor that is involved in regulation of organogenesis. Seems to be a regulator of SIX1, SIX6 and probably SIX5. Corepression of precursor cell proliferation in myoblasts by SIX1 is switched to coactivation through recruitment of EYA3 to the SIX1-DACH1 complex. Transcriptional activation seems also to involve association of CREBBP. Seems to act as a corepressor of SIX6 in regulating proliferation by directly repressing cyclin-dependent kinase inhibitors, including the p27Kip1 promoter. Inhibits TGF-beta signaling through interaction with SMAD4 and NCOR1. Binds to chromatin DNA via its DACHbox-N domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dac homolog; Dach1; dachshund family transcription factor 1; Dachshund homolog 1; FLJ10138; OTTHUMP00000040839; RP11-512J14.1
Gene Aliases: Dac; DACH; DACH1
UniProt ID: (Human) Q9UI36, (Mouse) Q9QYB2
Entrez Gene ID: (Human) 1602, (Rat) 306096, (Mouse) 13134
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support