Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Transcription factor that is involved in regulation of organogenesis. Seems to be a regulator of SIX1, SIX6 and probably SIX5. Corepression of precursor cell proliferation in myoblasts by SIX1 is switched to coactivation through recruitment of EYA3 to the SIX1-DACH1 complex. Transcriptional activation seems also to involve association of CREBBP. Seems to act as a corepressor of SIX6 in regulating proliferation by directly repressing cyclin-dependent kinase inhibitors, including the p27Kip1 promoter. Inhibits TGF-beta signaling through interaction with SMAD4 and NCOR1. Binds to chromatin DNA via its DACHbox-N domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dac homolog; Dach1; dachshund family transcription factor 1; Dachshund homolog 1; FLJ10138; OTTHUMP00000040839; RP11-512J14.1
Gene Aliases: Dac; DACH; DACH1
UniProt ID: (Human) Q9UI36, (Mouse) Q9QYB2
Entrez Gene ID: (Human) 1602, (Rat) 306096, (Mouse) 13134
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support