Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MGRAELLEGK MSTQDPSDLW SRSDGEAELL QDLGWYHGNL TRHAAEALLL SNGCDGSYLL RDSNETTGLY SLSVRAKDSV KHFHVEYTGY SFKFGFNEFS SLKDFVKHFA NQPLIGSETG TLMVLKHPYP RKVEEPSIYE SVRVHTAMQT GRTEDDLVPT APSLGTKEGY LTKQGGLVKT WKTRWFTLHR NELKYFKDQM SPEPIRILDL TECSAVQFDY SQERVNCFCL VFPFRTFYLC AKTGVEADEW IKILRWKLSQ IRKQLNQGEG TIRSRSFIFK
May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B lymphocyte adapter protein Bam32; B-cell adapter molecule of 32 kDa; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; hDAPP1
Gene Aliases: BAM32; DAPP1; HSPC066
UniProt ID: (Human) Q9UN19
Entrez Gene ID: (Human) 27071
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support