Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSAANPETP NSTISREAST QSSSAAASQG WVLPEGKIVP NTVFVGGIDA RMDETEIGSC FGRYGSVKEV KIITNRTGVS KGYGFVSFVN DVDVQKIVGS QIHFHGKKLK LGPAIRKQKL CARHVQPRPL VVNPPPPPQF QNVWRNPNTE TYLQPQITPN PVTQHVQAYS AYPHSPGQVI TGCQLLVYNY QEYPTYPDSA FQVTTGYQLP VYNYQPFPAY PRSPFQVTAG YQLPVYNYQA FPAYPNSPFQ VATGYQFPVY NYQPFPAYPS SPFQVTAGYQ LPVYNYQAFP AYPNSPFQVA TGYQFPVYNY QAFPAYPNSP VQVTTGYQLP VYNYQAFPAY PNSA (1-343 aa encoded by BC047617)
This gene is a member of the DAZ gene family and is a candidate for the human Y-chromosomal azoospermia factor (AZF). This RNA-binding protein is important for spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deleted in azoospermia protein 4
Gene Aliases: DAZ4; pDP1680; pDP1681
UniProt ID: (Human) Q86SG3
Entrez Gene ID: (Human) 57135
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support