Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PVGPIYPPGS TVLVEGGYDA GARFGAGATA GNIPPPPPGC PPNAAQLAVM QGANVLVTQR KGNFFMGGSD GGYTIW
DAZAP2 encodes a proline-rich protein which interacts with the deleted in azoospermia (DAZ) and the deleted in azoospermia-like gene through the DAZ-like repeats. This protein also interacts with the transforming growth factor-beta signaling molecule SARA (Smad anchor for receptor activation), eukaryotic initiation factor 4G, and an E3 ubiquitinase that regulates its stability in splicing factor containing nuclear speckles. The encoded protein may function in various biological and pathological processes including spermatogenesis, cell signaling and transcription regulation, formation of stress granules during translation arrest, RNA splicing, and pathogenesis of multiple myeloma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAZ-associated protein 2; deleted in azoospermia associated protein 2; Deleted in azoospermia-associated protein 2; proline-rich transcript in brain; Proline-rich transcript in brain protein; proline-rich transcript, brain-expressed protein
Gene Aliases: DAZAP2; KIAA0058; PRTB
UniProt ID: (Human) Q15038
Entrez Gene ID: (Human) 9802
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support