Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNWRFVELL YFLFIWGRIS VQPSHQEPAG TDQHVSKEFD WLISDRGPFH HSRSYLSFVE RHRQGFTTRY KIYREFARWK VRNTAIERRD LVRHPVPLMP EFQRSIRLLG RRPTTQQFID TIIKKYGTHL LISATLGGEE ALTMYMDKSR LDRKSGNATQ SVEALHQLAS SYFVDRDGTM RRLHEIQIST GAIKVTETRT GPLGCNSYDN LDSVSSVLLQ STESKLHLQG LQIIFPQYLQ EKFVQSALSY IMCNGEGEYL CQNSQCRCQC AEEFPQCNCP ITDIQIMEYT LANMAKSWAE AYKDLEN (1-320 aa encoded by BC021560)
This gene is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5' CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)); BMP/retinoic acid-inducible neural-specific protein (BRINP); BMP/retinoic acid-inducible neural-specific protein 1; BMP/retinoic acid-inducible neural-specific protein-1; bone morphogenetic protein/retinoic acid inducible neural-specific 1; bone morphogenic protein/retinoic acid inducible neural-specific 1; deleted in bladder cancer 1; deleted in bladder cancer chromosome region candidate 1; Deleted in bladder cancer protein 1; Deleted in bladder cancer protein 1 homolog
Gene Aliases: BRINP; BRINP1; DBC1; DBCCR1; FAM5A; IB3089A
UniProt ID: (Human) O60477, (Mouse) Q920P3
Entrez Gene ID: (Human) 1620, (Pig) 100625235, (Mouse) 56710
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support