Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: VKKQQPKKPE GTCPAAESRT RKVARLKAPF LKIEDESRKF RPFHHQFKSF PEISFLGPKD ASPFEAPTTL GSMHHTRESK DGEPSPRSAA HTMPRRKKG
This gene encodes a regulator of the CDC7-like 1 protein, a serine-threonine kinase which links cell cycle regulation to genome duplication. This protein localizes to the nucleus and its expression is cell cycle-regulated. Alternative splicing of this gene results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full length sequences have not been determined. [provided by RefSeq].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator of S phase kinase-like protein 1; activator of S-phase kinase-like protein 1; ASK-like protein 1; Chiffon homolog B; DBF4 homolog B; Dbf4-related factor 1; Protein DBF4 homolog B; zinc finger, DBF-type containing 1B
Gene Aliases: ASKL1; CHIFB; DBF4B; DRF1; ZDBF1B
UniProt ID: (Human) Q8NFT6
Entrez Gene ID: (Human) 80174
Molecular Function:
kinase activator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support