Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MSSKGSSTDG RTDLANGSLS SSPEEMSGAE EGRETSSGIE VEASDLSLSL TGDDGGPNRT STESRGTETE SSGEDKDSDS MEDTGHYSIN DENRVHDRSE EEEEEEEEEE EEQPRRRVQR KRANRDQDSS DDERALEDWV SSETSALPRP RWQALPALRE RELGSSARFV YEACGARVFV QRFRLQHGLE GHTGCVNTLH FNQRGTWLAS GSDDLKVVVW DWVRRQPVLD FESGHKSNVF QVRQGSIIAT ERIRHELEKS ELQHGLGADS ELL
This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DDB1- and CUL4-associated factor 8; WD repeat domain 42A; WD repeat-containing protein 42A
Gene Aliases: DCAF8; GAN2; H326; WDR42A
UniProt ID: (Human) Q5TAQ9
Entrez Gene ID: (Human) 50717
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support