Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSPLWWGFLL SCLGCKILPG AQGQFPRVCM TVDSLVNKEC CPRLGAESAN VCGSQQGRGQ CTEVRADTRP WSGPYILRNQ DDRELWPRKF FHRTCKCTGN FAGYNCGDCK FGWTGPNCER KKPPVIRQNI HSLSPQEREQ FLGALDLAKK RVHPDYVITT QHWLGLLGPN GTQPQFANCS VYDFFVWLHY YSVRDTLLGP GRPYRAIDFS HQGPAFVTWH RYHLLCLERD LQRLIGNESF ALPYWNFATG RNECDVCTDQ LFGAARPDDP TLISRNSRFS SWETVCDSLD DYNHLVTLCN GTYEGLLRRN QMGRNSMKLP TLKDIRDCLS LQKFDNPPFF QNSTFSFRNA LEGFDKADGT LDSQVMSLHN LVHSFLNGTN ALPHSAANDP IFVVLHSFTD AIFDEWMKRF NPPADAWPQE LAPIGHNRMY NMVPFFPPVT NEELFLTSDQ LGYSYAIDLP VSVEETPGWP TTLLVVMGTL VALVGLFVLL AFLQYRRLRK GYTPLMETHL SSKRYTEEA
The TYR-related proteins, TRP1 and TRP2, are also specifically expressed in melanocytes, and they likewise contribute to the synthesis of melanin within the melanosomes. The TRPs, including TYR, all share a similar transmembrane region, contain two metal-binding regions and a cysteinerich epidermal growth factor motif, and are localized in the melanosomal membrane. These proteins, however, have distinct catalytic activity, and they individually contribute to the biosynthesis of melanin biopolymers. The TRPs are believed to exist as a multi-enzyme complex, as these proteins form aggregates together, and the expression of TRP1 also helps stabilize TYR in melanocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DCT; dopachrome delta-isomerase; dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2); DT; L-dopachrome Delta-isomerase; L-dopachrome isomerase; L-dopachrome tautomerase; TRP-2; TRP2; tyrosinase related protein-2; Tyrosinase-related protein 2; tyrosine-related protein 2
Gene Aliases: DCT; TRP-2; TYRP2
UniProt ID: (Human) P40126
Entrez Gene ID: (Human) 1638
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support