Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 100%
The TYR-related proteins, TRP1 and TRP2, are also specifically expressed in melanocytes, and they likewise contribute to the synthesis of melanin within the melanosomes. The TRPs, including TYR, all share a similar transmembrane region, contain two metal-binding regions and a cysteinerich epidermal growth factor motif, and are localized in the melanosomal membrane. These proteins, however, have distinct catalytic activity, and they individually contribute to the biosynthesis of melanin biopolymers. The TRPs are believed to exist as a multi-enzyme complex, as these proteins form aggregates together, and the expression of TRP1 also helps stabilize TYR in melanocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DCT; DOPAchrome conversion factor; DOPAchrome isomerase; DOPAchrome oxidoreductase; L-dopachrome Delta-isomerase; L-dopachrome tautomerase; melanogenic enzyme; tyrosinase-related protein 2; trp2; SLATY locus protein; TRP-2; Tyrosinase-related protein 2; tyrosinase-related protein-2
Gene Aliases: Dct; DT; slaty; slt; TRP-2; TRP2; Tyrp-2; Tyrp2; zgc:109860
UniProt ID: (Mouse) P29812
Entrez Gene ID: (Mouse) 13190, (Zebrafish) 58074
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support