Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSVAGGEIR GDTGGEDTAA PGRFSFSPEP TLEDIRRLHA EFAAERDWEQ FHQPRNLLLA LVGEVGELAE LFQWKTDGEP GPQGWSPRER AALQEELSDV LIYLVALAAR CRVDLPLAVL SKMDINRRRY PAHLARSSSR KYTELPHGAI SEDQAVGPAD IPCDSTGQTS T (1-170 aa encoded by BC001344)
The protein encoded by this gene is dCTP pyrophosphatase, which converts dCTP to dCMP and inorganic pyrophosphate. The encoded protein also displays weak activity against dTTP and dATP, but none against dGTP. This protein may be responsible for eliminating excess dCTP after DNA synthesis and may prevent overmethylation of CpG islands. Three transcript variants, one protein-coding and the other two non-protein coding, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dCTP pyrophosphatase 1; dCTPase 1; Deoxycytidine-triphosphatase 1; RS21-C6; RS21C6; XTP3-transactivated gene A protein; XTP3-transactivated gene A protein homolog; XTP3-transactivated protein A
Gene Aliases: 2410015N17Rik; AI854235; CDA03; DCTPP1; RS21-C6; RS21C6; Tdrg-TL1; XTP3TPA
UniProt ID: (Human) Q9H773, (Rat) Q91VC0, (Mouse) Q9QY93
Entrez Gene ID: (Human) 79077, (Rat) 192252, (Mouse) 66422
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support