Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MELDFGHFDE RDKTSRNMRG SRMNGLPSPT HSAHCSFYRT RTLQALSNEK KAKKVRFYRN GDRYFKGIVY AVSSDRFRSF DALLADLTRS LSDNINLPQG VRYIYTIDGS RKIGSMDELE EGESYVCSSD NFFKKVEYTK NVNPNWSVNV KTSANMKAPQ SLASSNSAQA RENKDFVRPK LVTIIRSGVK PRKAVRVLLN KKTAHSFEQV LTDITEAIKL ETGVVKKLYT LDGKQVTCLH DFFGDDDVFI ACGPEKFRYA QDDFSLDENE CRVMKGNPSA TAGPKASPTP QKTSAKSPGP MRRSKSPADS ANGTSSSQLS TPKSKQSPIS TPTSPGSLRK HKDLYLPLSL DDSDSLGDSM
In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: doublecortex; Doublin; Lis-X; LIS-XD; Lissencephalin-X; Neuronal migration protein doublecortin; OTTHUMP00000023860; OTTHUMP00000216316
Gene Aliases: DBCN; DC; DCX; LISX; SCLH; XLIS
UniProt ID: (Human) O43602
Entrez Gene ID: (Human) 1641
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support