Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to DDB1.
Immunogen sequence: MSYNYVVTA QKPTAVNGCV TGHFTSAEDL NLLIAKNTRL EIYVVTAEGL RPVKEVGMYG KIAVMELFRP KGESKDLLFI LTAKYNACIL EYKQSGESID IITRAHGNVQ DRIGRPSETG IIGIIDPECR MIGLRLYDGL FKVIPLDRDN KELKAFNIRL EELHVIDVKF LYGCQAPTIC FVYQDPQGRH VKTYEVSLRE KEFNKGPWKQ ENVEAEASMV IAVPEPFGGA IIIGQESITY HNGDKYLAIA PPIIKQSTIV CHNRVDPNGS RYLLGDMEGR LFMLLLEKEE QMDGTVTLKD LRVELLGETS IAECLTYLDN GVVFVGSRLG DSQLVKLNVD SNEQGSYVVA METFTNLGPI VDMCVVDLER QGQGQLVTCS GAFKEGSLRI IRNGIGIHEH A (1-400 aa encoded by BC011686)
The DNA damage-binding protein from HeLa cells is associated with polypeptides of relative mass 124,000 (DDB1) and 41,000 (DDB2) as determined by SDS-polyacrylamide gels. To test whether the DNA-repair defect in the subset of XPE patients that lack DNA damage-binding activity is caused by a defect in DDB, Keeney et al. (1994) injected purified human DDB protein into XPE cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: damage specific DNA binding protein 1; damage-specific DNA binding protein 1, 127kDa; Damage-specific DNA-binding protein 1; damage-specific DNA-binding protein, DNA repair; damaged-DNA recognition protein 1; DDB p127 subunit; DDBa; DNA damage-binding protein 1; DNA damage-binding protein a; DNA repair protein; HBV X-associated protein 1; UV-damaged DNA-binding factor; UV-damaged DNA-binding protein 1; UV-DDB 1; XAP-1; xeroderma pigmento; Xeroderma pigmentosum group E-complementing protein; XPCe; XPE-BF; XPE-binding factor
Gene Aliases: 127kDa; AA408517; DDB1; DDBA; p127-Ddb1; UV-DDB1; XAP1; XPCE; XPE; XPE-BF
UniProt ID: (Human) Q16531, (Mouse) Q3U1J4
Entrez Gene ID: (Human) 1642, (Mouse) 13194, (Rat) 64470
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support