Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNYPGRGSPR SPEHNGRGGG GGAWELGSDA RPAFGGGVCC FEHLPGGDPD DGDVPLALLR GEPGLHLAPG TDDHNHHLAL DPCLSDENYD FSSAESGSSL RYYSEGESGG GGSSLSLHPP QQPPLVPTNS GGGGATGGSP GERKRTRLGG PAARHRYEVV TELGPEEVRW FYKEDKKTWK PFIGYDSLRI ELAFRTLLQT TGARPQGGDR DGDHVCSPTG PASSSGEDDD EDRACGFCQS TTGHEPEMVE LVNIEPVCVR GGLYEVDVTQ GECYPVYWNQ ADKIPVMRGQ WFIDGTWQPL EEEESNLIEQ EHLNCFRGQQ MQENFDIEVS KSIDGKDAVH SFKLSRNHVD WHSVDEVYLY SDATTSKIAR TVTQKLGFSK ASSSGTRLHR GYVEEATLED KPSQTTHIVF VVHGIGQKMD QGRIIKNTAM MREAARKIEE RHFSNHATHV EFLPVEWRSK LTLDGDTVDS ITPDKVRGLR DMLNSSAMDI MYYTSPLYRD ELVKGLQQEL NRLYSLFCSR NPDFEEKGGK VSIVSHSLGC VITYDIMTGW NPVRLYEQLL QKEEELPDER WMSYEERHLL DELYITKRRL KEIEERLHGL KASSMTQTPA LKFKVENFFC MGSPLAVFLA LRGIRPGNTG SQDHILPREI CNRLLNIFHP TDPVAYRLEP LILKRYSNIS PVQIHWYNTS NPLPYEHMKP SFLNPAKEPT SVSENEGIST IPSPVTSPVL SRRHYGESIT NIGKASILGA ASIGKGLGGM LFSRFGRSST TQSSETSKDS MEDEKKPVAS PSATTVGTQT LPHSSSGFLD SALELDHRID FELREGLVES RYWSAVTSHT AYWSSLDVAL FLLTFMYKHE HDDDAKPNLD PI
This gene is a member of the intracellular phospholipase A1 gene family. The protein encoded by this gene preferentially hydrolyzes phosphatidic acid. It is a cytosolic protein with some mitochondrial localization, and is thought to be involved in the regulation of mitochondrial dynamics. Overexpression of this gene causes fragmentation of the tubular structures in mitochondria, while depletion of the gene results in mitochondrial tubule elongation. Deletion of this gene in male mice caused fertility defects, resulting from disruption in the organization of the mitochondria during spermiogenesis. In humans, mutations in this gene have been associated with hereditary spastic paraplegia (HSP), also known as Strumpell-Lorrain disease, or, familial spastic paraparesis (FSP). This inherited disorder is characterized by progressive weakness and spasticity of the legs. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DDHD domain-containing protein 1; PA-PLA1; Phosphatidic acid-preferring phospholipase A1 homolog; phosphatidic acid-preferring phospholipase A1-like protein; Phospholipase DDHD1; Phospholipid sn-1 acylhydrolase; spastic paraplegia 28 (autosomal recessive)
Gene Aliases: DDHD1; KIAA1705; PA-PLA1; PAPLA1; SPG28
UniProt ID: (Human) Q8NEL9
Entrez Gene ID: (Human) 80821
Molecular Function:
hydrolase
lipase
phospholipase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support