Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: WNLSPDAENS REILSRCCAL EPSLHGACNI REKVGLRPYR PGVRLQTELL ARDGQRLPVV HHYGHGSGGI SVHWGTALEA ARLVSECVHA LRTPIPKSNL
DDO is a peroxisomal flavoprotein that catalyzes the oxidative deamination of D-aspartate and N-methyl D-aspartate. Flavin adenine dinucleotide or 6-hydroxyflavin adenine dinucleotide can serve as the cofactor in this reaction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aspartic oxidase; D-aspartate oxidase; D-aspartate oxidase, DDO; DASOX
Gene Aliases: DASOX; DDO; DDO-1; DDO-2
UniProt ID: (Human) Q99489
Entrez Gene ID: (Human) 8528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support