Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
DDR2 is a member of a subclass of RTKs and contains a distinct extracellular region encompassing a factor VIII-like domain. DDR1 and DDR2 have been shown to be potently inhibited by Gleevec. DDR2 required for normal bone development. Regulates osteoblast differentiation and chondrocyte maturation via a signaling pathway that involves MAP kinases and leads to the activation of the transcription factor RUNX2. Regulates remodeling of the extracellular matrix by up-regulation of the collagenases MMP1, MMP2 and MMP13, and thereby facilitates cell migration and tumor cell invasion. Promotes fibroblast migration and proliferation, and thereby contributes to cutaneous wound healing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD167 antigen-like family member B; CD167b; CD167b antigen; CD_antigen: CD167b; cell migration-inducing protein 20; Discoidin domain receptor 2; discoidin domain receptor family, member 2; Discoidin domain-containing receptor 2; Discoidin domain-containing receptor tyrosine kinase 2; hydroxyaryl-protein; hydroxyaryl-protein kinase; migration-inducing gene 16 protein; neurotrophic tyrosine kinase receptor related 3; Neurotrophic tyrosine kinase, receptor-related 3; Receptor protein-tyrosine kinase TKT; RP11-572K18.1; tyrosine-protein kinase TYRO 10; Tyrosine-protein kinase TYRO10; tyrosylprotein kinase
Gene Aliases: AW495251; DDR2; MIG20a; NTRKR3; TKT; TYRO10
UniProt ID: (Human) Q16832, (Mouse) Q62371
Entrez Gene ID: (Human) 4921, (Mouse) 18214, (Rat) 685781
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support