Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
ATP-dependent RNA helicase involved in mRNA export from the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase DDX19A; DDX19-like protein; DEAD (Asp-Glu-Ala-As) box polypeptide 19A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19A; DEAD box protein 19A; DEAD box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19; eukaryotic translation initiation factor 4A related sequence 1; Eukaryotic translation initiation factor 4A-related sequence 1; mDEAD5; RNA helicase; zinc responsive protein ZD10B
Gene Aliases: DBP5; Ddx19; DDX19-DDX19L; DDX19A; DDX19L; Eif4a-rs1; ZD10B
UniProt ID: (Human) Q9NUU7, (Mouse) Q61655
Entrez Gene ID: (Human) 55308, (Mouse) 13680, (Rat) 292022
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support