Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
ATP-dependent RNA helicase involved in mRNA export from the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase DDX19A; DDX19-like protein; DEAD (Asp-Glu-Ala-As) box polypeptide 19A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19A; DEAD box protein 19A; RNA helicase
Gene Aliases: DDX19-DDX19L; DDX19A; DDX19L
UniProt ID: (Human) Q9NUU7
Entrez Gene ID: (Human) 55308
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support