Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: ASTEPEQLGS VPTDESAITQ MCEVTGGRSY CVRTQRMLNQ CLESLVQKVQ
Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
DDX26B is a protein coding gene. An important paralog of this gene is INTS6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B; Integrator complex subunit 6-like; Protein DDX26B
Gene Aliases: DDX26B; INTS6L
UniProt ID: (Human) Q5JSJ4
Entrez Gene ID: (Human) 203522
Molecular Function:
RNA metabolism protein
RNA helicase
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support