Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: DLERGCHLLV ATPGRLVDMM ERGKIGLDFC KYLVLDEADR MLDMGFEPQI RRIVEQDTMP PKGVRHTMMF SATFPKEIQM LARDFLDEYI FLAVGRVGST SENITQKVVW VEESDKRSFL LDLLNATGKD SLTLVFVETK KGADSLEDFL YHEGYACTSI HGDRSQRDRE EALHQFRSGK SPILVATAVA ARGLDISNVK HVINFDLPSD IEEYVHRIGR TGRVGNLGLA TSFFNERNIN ITKDLLDLLV EAKQEVPSWL ENMAYEHHYK GSSRGRSKSS RFSGGFGARD YRQSSGASSS SFSSSRASSS RSGGGGHGSS RGFGGGGYGG FYNSDGYGGN YNSQGVDWWG N
The protein encoded by this gene is a member of the large DEAD-box protein family, that is defined by the presence of the conserved Asp-Glu-Ala-Asp (DEAD) motif, and has ATP-dependent RNA helicase activity. This protein has been reported to display a high level of RNA-independent ATPase activity, and unlike most DEAD-box helicases, the ATPase activity is thought to be stimulated by both RNA and DNA. This protein has multiple conserved domains and is thought to play roles in both the nucleus and cytoplasm. Nuclear roles include transcriptional regulation, mRNP assembly, pre-mRNA splicing, and mRNA export. In the cytoplasm, this protein is thought to be involved in translation, cellular signaling, and viral replication. Misregulation of this gene has been implicated in tumorigenesis. This gene has a paralog located in the nonrecombining region of the Y chromosome. Pseudogenes sharing similarity to both this gene and the DDX3Y paralog are found on chromosome 4 and the X chromosome. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase DDX3X; CAP-Rf; DBX; DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked; DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked; DEAD box protein 3, X-chromosomal; DEAD box, X isoform; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, X-linked; DEAD/H box-3; Helicase-like protein 2; HLP2
Gene Aliases: CAP-Rf; DBX; DDX14; DDX3; DDX3X; HLP2; MRX102
UniProt ID: (Human) O00571
Entrez Gene ID: (Human) 1654, (Rat) 317335
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support