Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Polyclonal anti-DDX3Y antibodies (14041-1-AP) are produced by immunizing animals with C-terminus of DDX3Y and detects a 73 kDa band. As DDX3Y has a structural homolog with DDX3X, the antibody can recognize both.
Immunogen sequence: LERGCHLLV ATPGRLVDMM ERGKIGLDFC KYLVLDEADR MLDMGFEPQI RRIVEQDTMP PKGVRHTMMF SATFPKEIQM LARDFLDEYI FLAVGRVGST SENITQKVVW VEDLDKRSFL LDILGATGSD SLTLVFVETK KGADSLEDFL YHEGYACTSI HGDRSQRDRE EALHQFRSGK SPILVATAVA ARGLDISNVR HVINFDLPSD IEEYVHRIGR TGRVGNLGLA TSFFNEKNMN ITKDLLDLLV EAKQEVPSWL ENMAYEHHYK GGSRGRSKSN RFSGGFGARD YRQSSGSSSS GFGASRGSSS RSGGGGYGNS RGFGGGGYGG FYNSDGYGGN YNSQGVDWWG N (311-660 aa encoded by BC034942)
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases which are implicated in a number of cellular processes involving RNA binding and alteration of RNA secondary structure. DDX3Y is one member of DEAD box RNA helicases, encode by the gene locate on the chromosome Y in the azoospermia factor A region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase DDX3Y; D1Pas1 related sequence 1; D1Pas1-related sequence 1; DEAD (Asp-Glu-Ala-Asp) box helicase 3, Y-linked; DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide, Y chromosome; DEAD box protein 3, Y-chromosomal; DEAD-box RNA helicase DEAD2; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, Y chromosome; mDEAD2; uncharacterized protein LOC364073
Gene Aliases: 8030469F12Rik; D1Pas1-rs1; DBY; DDX3Y; Dead2; RGD1309586
UniProt ID: (Human) O15523, (Mouse) Q62095
Entrez Gene ID: (Human) 8653, (Rat) 364073, (Mouse) 26900
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support