Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MSHHGGAPKA STWVVASRRS STVSRAPERR PAEELNRTGP EGYSVGRGGR WRGTSRPPED VAAGHEELPL CFALKSHFVG AVIGRGGSKI KNIQSTTNTT IQIIQEQPES LVKIFGSKAM QTKAKAVIDN FVKKLEENYN SECGIDTAFQ PSVGKDGSTD NNVVAGDRPL IDWDQIREEG LKWQKTKWAD LPPIKKNFYK ESTATSAMSK VEADSWRKEN FNITWDDLKD GEKRPIPNPT CTFDDAFQCY PEVMENIKKA GFQKPTPIQS QAWPIVLQGI DLIGVAQTGT GKTLCYLMPG FIHLVLQPSL KGQRNRPGML VLTPTRELAL QVEGECCKYS YKGLRSVCVY GGGNRDEQIE ELKKGVDIII ATPGRLNDLQ MSNFVNLKNI TYLVLDEADK MLDMGFEPQI MKILLDVRPD RQTVMTSATW PHSVHRLAQS YLKEPMIVYV GTLDLVAVSS VKQNIIVTTE EEKWSHMQTF LQSMSSTDKV IVFVSRKAVA DHLSSDLILG NISVESLHGD REQRDREKAL ENFKTGKVRI LIATDLASRG LDVHDVTHVY NFDFPRNIEE YVHRIGRTGR AGRTGVSITT LTRNDWRVAS ELINILERAN QSIPEELVSM AERFKAHQQK REMERKMERP QGRPKKFH
The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 13; CT13; DEAD (Asp-Glu-Ala-Asp) box polypeptide 43; DEAD box protein 43; DEAD box protein HAGE; DEAD-box protein 43; Helical antigen; Probable ATP-dependent RNA helicase DDX43
Gene Aliases: CT13; DDX43; HAGE
UniProt ID: (Human) Q9NXZ2
Entrez Gene ID: (Human) 55510
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support