Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: IKEELLHSEK LKTYFEDNPR DLQLLRHDLP LHPAVVKPHL GHVPDYLVPP ALRGLVRPHK KRKKLSSSCR KAKRAKSQNP LRSFKHKGKK FRPTAKPS
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 61-kd nucleolar helicase; ATP-dependent 61 kDa nucleolar RNA helicase; DEAD (Asp-Glu-Ala-Asp) box helicase 56; DEAD (Asp-Glu-Ala-Asp) box polypeptide 56; DEAD box protein 21; DEAD box protein 56; DEAD-box RNA helicase; nucleolar helicase of 61 kDa; nucleolar RNA helicase homolog; Probable ATP-dependent RNA helicase DDX56; putative nucleolar RNA helicase; RNA helicase II/Gu
Gene Aliases: 2600001H07Rik; D11Ertd619e; DDX21; DDX26; DDX56; NOH61; RH-II/Gu
UniProt ID: (Human) Q9NY93, (Mouse) Q9D0R4
Entrez Gene ID: (Human) 54606, (Rat) 289780, (Mouse) 52513
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support