Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Dec; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix family, member e40; bHLHb2; bHLHe40; Class B basic helix-loop-helix protein 2; Class E basic helix-loop-helix protein 40; DEC1; differentially expressed in chondrocytes 1; Differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; Enhancer-of-split and hairy-related protein 2; FLJ99214; SHARP-2; Stimulated by retinoic acid gene 13 protein
Gene Aliases: BHLHB2; BHLHE40; DEC1; HLHB2; SHARP-2; SHARP2; STRA13; Stra14
UniProt ID: (Human) O14503
Entrez Gene ID: (Human) 8553
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support