Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DEDD encodes a protein that contains a death effector domain (DED). DED is a protein-protein interaction domain shared by adaptors, regulators and executors of the programmed cell death pathway. Overexpression of this gene was shown to induce weak apoptosis. Upon stimulation, this protein was found to translocate from cytoplasm to nucleus and colocalize with UBTF, a basal factor required for RNA polymerase I transcription, in the nucleolus. At least three transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Death effector domain-containing protein; Death effector domain-containing testicular molecule; DEDPro1; FLDED-1; tumor necrosis factor (ligand) superfamily, member 19
Gene Aliases: CASP8IP1; DEDD; DEDD1; DEDPRO1; DEFT; FLDED1; KE05
UniProt ID: (Human) O75618, (Mouse) Q9Z1L3, (Rat) Q9Z2K0
Entrez Gene ID: (Human) 9191, (Mouse) 21945, (Rat) 83631
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support