FIGURE: 1 / 1
Sequence of this protein is as follows: MRTLTILTAV LLVALQAKAE PLQAEDDPLQ AKAYEADAQE QRGANDQDFA VSFAEDASSS LRALGSTRAF TCHCRRSCYS TEYSYGTCTV MGINHRFCCL
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes appear to be clustered on chromosome 8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: defensin 6; Defensin, alpha 6; defensin, alpha 6, Paneth cell-specific; Defensin-6
Gene Aliases: DEF6; DEFA6; HD-6
UniProt ID: (Human) Q01524
Entrez Gene ID: (Human) 1671
Molecular Function:
antimicrobial response protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support