Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: Beta-defensin 110.
Defensins form a family of antimicrobial and cytotoxic peptides made by neutrophils. Defensins are short, processed peptide molecules that are classified by structure into three groups: alpha-defensins, beta-defensins and theta-defensins. All beta-defensin genes are densely clustered in four to five syntenic chromosomal regions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-defensin 10; Beta-defensin 11; Beta-defensin 110; Beta-defensin 111; DEFB-10; DEFB-11; defensin, beta 10; Defensin, beta 110; defensin, beta 110 locus; Defensin, beta 111
Gene Aliases: DEFB-10; DEFB-11; DEFB10; DEFB11; DEFB110; DEFB111
UniProt ID: (Human) Q30KQ9
Entrez Gene ID: (Human) 245913
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support