Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Guanine nucleotide exchange factor (GEF) which may activate RAB9A and RAB9B. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DENN domain-containing protein 2C; DENN/MADD domain containing 2C; RP5-1156J9.1
Gene Aliases: DENND2C; dJ1156J9.1
UniProt ID: (Human) Q68D51
Entrez Gene ID: (Human) 163259
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support