Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MATVQEKAAA LNLSALHSPA HRPPGFSVAQ KPFGATYVWS SIINTLQTQV EVKKRRHRLK RHNDCFVGSE AVDVIFSHLI QNKYFGDVDI PRAKVVRVCQ ALMDYKVFEA VPTKVFGKDK KPTFEDSSCS LYRFTTIPNQ DSQLGKENKL YSPARYADAL FKSSDIRSAS LEDLWENLSL KPANSPHVNI SATLSPQVIN EVWQEETIGR LLQLVDLPLL DSLLKQQEAV PKIPQPKRQS TMVNSSNYLD RGILKAYSDS QEDEWLSAAI DCLEYLPDQM VVEISRSFPE QPDRTDLVKE LLFDAIGRYY SSREPLLNHL SDVHNGIAEL LVNGKTEIAL EATQLLLKLL DFQNREEFRR LLYFMAVAAN PSEFKLQKES DNRMVVKRIF SKAIVDNKNL SKGKTDLLVL FLMDHQKDVF KIPGTLHKIV SVKLMAIQNG RDPNRDAGYI YCQRIDQRDY SNNIEKTTKD ELLNLLKTLD EDSKLSAKEK KKLLGQFYKC HPDIFIEHFG D
DEPDC7 gene ontology annotations related to this gene include intracellular signal transduction; regulation of small GTPase mediated signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEP domain-containing protein 7; dJ85M6.4 (novel 58.3 KDA protein); novel 58.3 KDA protein; Protein TR2/D15
Gene Aliases: AV216087; DEPDC7; dJ85M6.4; RGD1309720; TR2
UniProt ID: (Human) Q8NCU9, (Mouse) Q91WS7, (Rat) Q4QR86
Entrez Gene ID: (Human) 91614, (Mouse) 211896, (Rat) 295971
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support