Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: NPSEFKLQKE SDNRMVVKRI FSKAIVDNKN LSKGKTDLLV LFLMDHQKDV FKIPGTLHKI VSVKLMAIQN GRDPNRDAGY IYCQRIDQRD YSNNTEKTTK DELLNLL
DEPDC7 gene ontology annotations related to this gene include intracellular signal transduction; regulation of small GTPase mediated signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEP domain-containing protein 7; dJ85M6.4 (novel 58.3 KDA protein); novel 58.3 KDA protein; Protein TR2/D15
Gene Aliases: DEPDC7; dJ85M6.4; TR2
UniProt ID: (Human) Q8NCU9
Entrez Gene ID: (Human) 91614
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support