Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C17orf81 belongs to the ELP5 family. C17orf81 may be involved in TP53-mediated transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dermal papilla derived protein 6; Dermal papilla-derived protein 6; Dermal papilla-derived protein 6 homolog; Elongator complex protein 5; retinoic acid induced 12; Retinoic acid-induced protein 12; S-phase 2 protein
Gene Aliases: C17orf81; DERP6; ELP5; HSPC002; MST071; MSTP071; Rai12
UniProt ID: (Human) Q8TE02, (Mouse) Q99L85, (Rat) Q6IUP3
Entrez Gene ID: (Human) 23587, (Mouse) 54351, (Rat) 287446
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support