Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLGARVAAH LDALGPLVPY VPPPLLPSMF YVGLFFVNVL ILYYAFLMEY IVLNVGLVFL PEDMDQALVD LGVLSDPGSG LYDADSELDV FDAYLE (1-95 aa encoded by BC001083)
DEXI (dexamethasone-induced protein), also known as MYLE, is a 95 amino acid protein belonging to the DEXI family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dexamethasone-induced protein; dexamethasone-induced transcript; Protein MYLE
Gene Aliases: 1810029J14Rik; AI836170; AW413143; D16Bwg0586e; DEXI; MYLE; RGD1564938
UniProt ID: (Human) O95424, (Mouse) Q9WUQ7
Entrez Gene ID: (Human) 28955, (Mouse) 58239, (Rat) 497857
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support