Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Developmentally regulated transcription factor AP-1 binds and recognizes the enhancer DNA sequence: 5'-TGA[CG]TCA-3'. May play a role in the function or determination of a particular subset of cells in the developing embryo. It is able to carry out its function either independently of or in conjunction with Jra.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activator protein 1; AP-1; CG33956-PA; CG33956-PB; CG33956-PD; CG33956-PF; CG33956-PG; Dfos; Dfos/kayak; dorsal holes D; Fos; Fos-related antigen; FRA; kay; kay Fra CG15509; kay-PA; kay-PB; kay-PD; kay-PF; kay-PG; SD04477p; Transcription factor kayak, isoforms A/B/F; Transcription factor kayak, isoforms D/sro; Transcription factor kayak,isoforms A/B/F
Gene Aliases: AP-1; AP1; c-Fos; CG15507; CG15509; CG33956; D-Fos; D-fos/kay; dAP-1; DFOS; DFra; dlhD; DM3; DM30; Dmel\CG33956; Dmel_CG33956; fbz; FOS; Fra; KAY
UniProt ID: (Fruit fly) P21525
Entrez Gene ID: (Fruit fly) 3772082
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support