Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MALRWGIVS VGLISSDFTA VLQTLPRSEH QVVAVAARDL SRAKEFAQKH DIPKAYGSYE ELAKDPSVEV AYIGTQHPQH KAAVMLCLAA GKAVLCEKPT GVNAAEVREM VAEARSRALF LMEAIWTRFF PASEALRSVL AQGTLGDLRV ARAEFGKNLI HVPRAVDRAQ AGGALLDIGI YCVQFTSMVF GGQKPEKISV VGRRHETGVD DTVTVLLQYP GEVHGSFTCS ITVQLSNTAS VSGTKGMVQL LNPCWCPTEL VVKGEHKEFP LPPVPKDCNF DNGAGMSYEA KHVWECLRKG MKESPVIPLS ESELLADILE EVRKAIGVTF PQDKR (1-334 aa encoded by BC032730)
This gene encodes an enzyme that belongs to the family of dihydrodiol dehydrogenases, which exist in multiple forms in mammalian tissues and are involved in the metabolism of xenobiotics and sugars. These enzymes catalyze the NADP1-linked oxidation of transdihydrodiols of aromatic hydrocarbons to corresponding catechols. This enzyme is a dimeric dihydrodiol dehydrogenase, and it differs from monomeric dihydrodiol dehydrogenases in its high substrate specificity for trans-dihydrodiols of aromatic hydrocarbons in the oxidative direction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-deoxyglucosone reductase; D-xylose 1-dehydrogenase; D-xylose-NADP dehydrogenase; DDH; dihydrodiol dehydrogenase (dimeric); Dimeric dihydrodiol dehydrogenase; Hum2DD; Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; xylose dehydrogenase 1
Gene Aliases: 1300018L09Rik; 2DD; AA591799; DHDH; HUM2DD; Xld-1; Xld1
UniProt ID: (Human) Q9UQ10, (Mouse) Q9DBB8
Entrez Gene ID: (Human) 27294, (Rat) 691002, (Mouse) 71755
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support