Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TDILVRHQSF VLVGETGSGK TTQIPQWCVE YMRSLPGPKR GVACTQPRRV AAMSVAQRVA DEMDVMLGQE VGYSIRFEDC SSAKTILKYM TDGMLLREAM NDPLLERYGV IILDEAHERT LATDILMGVL KEVVRQRSDL KVIVMSATLD AGKFQIYFDN CPLLTIPGRT HPVEIFYTPE PERDYLEAAI RTVIQIHMCE EEEGDLLLFL TGQEEIDEAC KRIKREVDDL GPEVGDIKII PLYSTLPPQQ QQRIFEPPPP KKQNGAIGRK VVVSTNIAET SLTIDGVVFV IDPGFAKQKV YNPRIRVESL LVTAISKASA QQRAGRAGRT RPGKCFRLYT EKAYKTEMQD NTYPEILRSN LGSVVLQLKK L
DHX15 (DEAH-Box Helicase 15) is a Protein Coding gene. Among its related pathways are Gene Expression and mRNA Splicing - Major Pathway. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and helicase activity. An important paralog of this gene is DHX8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase #46; ATP-dependent RNA helicase DHX15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15 (RNA helicase A); DEAD/H box-15; DEAH (Asp-Glu-Ala-His) box helicase 15; DEAH (Asp-Glu-Ala-His) box polypeptide 15; DEAH box protein 15; hPrp43; pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; RNA helicase 2; Splicing factor Prp43
Gene Aliases: DBP1; DDX15; DEAH9; DHX15; HRH2; mDEAH9; PRP43; PRPF43; PrPp43p
UniProt ID: (Human) O43143, (Mouse) O35286
Entrez Gene ID: (Human) 1665, (Rat) 289693, (Mouse) 13204
Molecular Function:
RNA helicase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support