Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody reacts with 150 kDa DHX57 protein.
Immunogen sequence: PLEYAGLAS VEPYVPEFTV SPFAVQKLSR YGFNTERCQA VLRMCDGDVG ASLEHLLTQC FSETFGERMK ISEAVNQISL DECMEQRQEE AFALKSICGE KFIERIQNRV WTIGLELEYL TSRFRKSKPK ESTKNVQENS LEICKFYLKG NCKFGSKCRF KHEVPPNQIV GRIERSVDDS HLNAIEDASF LYELEIRFSK DHKYPYQAPL VAFYSTNENL PLACRLHISE FLYDKALTFA ETSEPVVYSL ITLLEEESEI VKLLTNTHHK YSDPPVNFLP VPSRTRINNP ACHKTVIPNN SFVSNQIPEV EKASESEESD EDDGPAPVIV ENESYVNLK (163-500 aa encoded by BC131534)
Probable ATP-binding RNA helicase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57; DEAH box protein 57; DEAH-box RNA/DNA helicase AAM73547; Putative ATP-dependent RNA helicase DHX57
Gene Aliases: DDX57; DHX57
UniProt ID: (Human) Q6P158
Entrez Gene ID: (Human) 90957
Molecular Function:
RNA helicase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support