Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPKVKSGAI GRRRGRQEQR RELKSAGGLM FNTGIGQHIL KNPLIINSII DKAALRPTDV VLEVGPGTGN MTVKLLEKAK KVVACELDPR LVAELHKRVQ GTPVASKLQV LVGDVLKTDL PFFDTCVANL PYQISSPFVF KLLLHRPFFR CAILMFQREF ALRLVAKPGD KLYCRLSINT QLLARVDHLM KVGKNNFRPP PKVESSVVRI EPKNPPPPIN FQEWDGLVRI TFVRKNKTLS AAFKSSAVQQ LLEKNYRIHC SVHNIVSAAV YPVKQI (1-275 aa encoded by B C002841)
DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3'-end of 18S rRNA in the 40S particle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; 18S rRNA dimethylase; DIM1 dimethyladenosine transferase 1 homolog; DIM1 dimethyladenosine transferase 1-like; dimethyladenosine transferase; Probable 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; Probable 18S rRNA dimethylase; Probable dimethyladenosine transferase; Probable S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase; S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase
Gene Aliases: 1500031M22Rik; AV111347; DIM1; DIMT1; DIMT1L; HSA9761; HUSSY-05; HUSSY5; RGD1311752
UniProt ID: (Human) Q9UNQ2, (Mouse) Q9D0D4
Entrez Gene ID: (Human) 27292, (Mouse) 66254, (Rat) 294718
Molecular Function:
RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support