Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
The protein encoded by this gene belongs to the iodothyronine deiodinase family. It activates thyroid hormone by converting the prohormone thyroxine by outer ring deiodination to bioactive 3,3',5-triiodothyronine residues encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence, which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5DII; deiodinase-2; deiodonase-2; DIOII; thyroxine deiodinase; thyroxine deiodinase, type II; thyroxine type II; Type 2 DI; type 2 iodothyronine deiodinase; Type II iodothyronine deiodinase; Type-II 5'-deiodinase; type-II 5'deiodinase
Gene Aliases: 5DII; D2; DIO2; DIOII; ITDI2; SelY; TXDI2
UniProt ID: (Human) Q92813
Entrez Gene ID: (Human) 1734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support