Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Required for the regulation of cell proliferation in response to extracellular signals mediated by an early endosomal compartment. Links Rab5 to nuclear signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adapter protein containing PH domain, PTB domain and leucine zipper motif 2; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2; DCC-interacting protein 13-beta; DIP13 beta; Dip13-beta; Dip3 beta
Gene Aliases: APPL2; DIP13B; Dip3b; RGD1563028
UniProt ID: (Human) Q8NEU8, (Rat) B4F779, (Mouse) Q8K3G9
Entrez Gene ID: (Human) 55198, (Rat) 362860, (Mouse) 216190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support