Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPEQSNDYRV VVFGAGGVGK SSLVLRFVKG TFRDTYIPTI EDTYRQVISC DKSVCTLQIT DTTGSHQFPA MQRLSISKGH AFILVFSVTS KQSLEELGPI YKLIVQIKGS VEDIPVMLVG NKCDETQREV DTREAQAVAQ EWKCAFMETS AKMNYNVKEL FQELLTLETR RNMSLNIDGK RSGKQKRTDR VKGKCTLM
DIRAS1 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DIRAS family GTP binding RAS like 1; DIRAS family, GTP-binding RAS-like 1; Distinct subgroup of the Ras family member 1; GTP-binding protein Di-Ras1; Ras-related inhibitor of cell growth; Rig; Small GTP-binding tumor suppressor 1
Gene Aliases: Di-Ras1; DIRAS1; GBTS1; RIG
UniProt ID: (Human) O95057
Entrez Gene ID: (Human) 148252
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support