Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MMALGAAGAT RVSVAMVAAA LGGHPLLGVS ATLNSVLNSN AIKNLPPPLG GAAGHPGSAV SAAPGILYPG GNKYQTIDNY QPYPCAEDEE CGTDEYCASP TRGGDAGVQI CLACRKRRKR CMRHAMCCPG NYCKNGICVS SDQNHFRGEI EETITESFGN DHSTLDGYSR RTTLSSKMYH TTGQEGSVCL RSSDCASGLC CARHFWSKIC KPVLKGGQVC TKHRRKGSHG LEIFQRCYCG EGLSCRIQKD HHQASNSSRL HTCQRH
DKK-1 is a member of the DKK protein family which also includes DKK-2, DKK-3 and DKK-4. DKK-1 was originally identified as a Xenopus head-forming molecule that behaves as an antagonist for Wnt signaling. Subsequent studies have shown that DKK-1 and DKK-4 play important regulatory roles in the Wnt/beta-catenin signaling pathway by forming inhibitory complexes with LDL receptor-related proteins 5 and 6 (LRP5 and LRP6), which are essential components of the Wnt/beta-catenin signaling system. LRP5 and LRP6 are single-pass transmembrane proteins that appear to act as co-receptors for Wnt ligands involved in the Wnt/beta-catenin signaling cascade. It has been suggested that by inhibiting Wnt/beta-catenin signaling, which is essential for posterior patterning in vertebrates, DKK-1 permits anterior development. This notion is supported by the finding that mice deficient of DKK-1 expression lack head formation and die during embryogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dickkopf 1 homolog; dickkopf related protein-1; Dickkopf-1; dickkopf-1 like; dickkopf-like protein 1; Dickkopf-related protein 1; Dickkopf1; hDkk-1; hDkk1; SK
Gene Aliases: DKK-1; DKK1; SK; UNQ492/PRO1008
UniProt ID: (Human) O94907
Entrez Gene ID: (Human) 22943
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support